Learn More
Bioworld Hepatitis B Virus Core Delta (Recombinant), 100 μg
Supplier:  Bioworld 220601021
Encompass_Preferred
Recombinant Hepatitis B Virus Core (HBcAg) Source: E.coli Product Description: E.coli derived recombinant. The protein contains the HBV core immunodominant region. Physical appearance: Sterile Filtered colourless liquid formulation Purification Metod: Purified by proprietary chromatographic technique. Purity: Protein is >90% pure as determined by 10% PAGE (coomassie staining). Formulation: 1mg/ml in 50% glycerol, 7.5mM phosphate buffer pH 7.2, 75mM NaCl. Storage: Protein is shipped at ambient temperature. Upon arrival, Store at -200C. Stability: Five years frozen. One month in solution at room temperature. Specificity: Immunoreactive with sera HBV-infected individuals. Applications: Antigen in ELISA and Western blots, excellent antigen for detection of HBV with minimal specificity problems. Usage: BioWORLD's products are furnished for LABORATORY RESEARCH USE ONLY. The product may not be used as drugs, agricultural or pesticidal products, food additives or household chemicals.
Catalog No. 50-253-203