Learn More
Bioworld Hepatitis B Virus HBe (Recombinant)   , 100 μg
Supplier:  Bioworld 220601041
Encompass_Preferred
Recombinant Hepatitis B Virus HBe Source: E.coli Product Description: E.coli derived recombinant. The protein contains the Hepatitis B Virus HBe immunodominant region. Purification Method: Purified by proprietary chromatographic technique. Purity: Protein is >95% pure as determined by 10% PAGE (coomassie staining). Protein Conc: 1mg/ml, 25mM Tris-HCl pH 8.0, 1.5 M urea and 50% glycerol. Storage: Protein is shipped at ambient temperature. Upon arrival, Store at -200C. Stability: Five years frozen. One month in solution at room temperature Specificity: Immunoreactive with sera HBV-infected individuals. Applications: Antigen in ELISA and Western blots, excellent antigen for detection of HBV with minimal specificity problems. Usage: BioWORLD\\\'s products are furnished for LABORATORY RESEARCH USE ONLY. The product may not be used as drugs, agricultural or pesticidal products, food additives or household chemicals.
Catalog No. 50-253-205
 Restricted Item