Learn More
Diagnostic Biosystems HEPATOCYTE Hepatocyte Specific Antigen (OCH1E5)
Catalog No. 502407598
Encompass_Preferred
Hepatoblastoma is the most common primary tumor of the liver in children. The use of specific hepatocyte markers and also of α fetoprotein or carcinoembryonic antigen are useful for the identification of normal and malignant fetal hepatocytes. This antibody recognizes an uncharacterized antigen present in both adults and fetal normal hepatocytes to produce a distinct granular cytoplasmic staining. This antibody stains majority of hepatocellular carcinomas.
Catalog No. 50-240-7598 Supplier Diagnostic Biosystems Supplier No. PDM162
Hepatoblastoma is the most common primary tumor of the liver in children. The use of specific hepatocyte markers and also of α fetoprotein or carcinoembryonic antigen are useful for the identification of normal and malignant fetal hepatocytes. This antibody recognizes an uncharacterized antigen present in both adults and fetal normal hepatocytes to produce a distinct granular cytoplasmic staining. This antibody stains majority of hepatocellular carcinomas.