Learn More
Sigma Aldrich Fine Chemicals Biosciences Hepatocyte Growth factor h
Supplier:  Sigma Aldrich Fine Chemicals Biosciences H96615UG
Encompass
Recombinant human hepatocyte growth factor (HGf) is produced from the 728 amino acid residue variant of the human pre-pro-HGf protein 6 and expressed in a mouse myeloma cell line NSO.The HGf gene is mapped to human chromosome 7q21.1. The encoded protein is a mesenchymal-fibroblast derived cytokine.
Catalog No. 50-290-4788