Learn More
Sigma Aldrich Fine Chemicals Biosciences Hepatocyte Growth factor h25UG
Catalog No. 502914513
Encompass
Recombinant human hepatocyte growth factor (HGf) is produced from the 728 amino acid residue variant of the human pre-pro-HGf protein 6 and expressed in a mouse myeloma cell line NSO.The HGf gene is mapped to human chromosome 7q21.1. The encoded protein is a mesenchymal-fibroblast derived cytokine.
Catalog No. 50-291-4513 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H966125UG
Recombinant human hepatocyte growth factor (HGf) is produced from the 728 amino acid residue variant of the human pre-pro-HGf protein 6 and expressed in a mouse myeloma cell line NSO.The HGf gene is mapped to human chromosome 7q21.1. The encoded protein is a mesenchymal-fibroblast derived cytokine.