Learn More
Medchemexpress LLC Hepatocyte growth factor (HGF) protein, cynomolgus, recombinant (HEK293) | >95.0% | 100 UG
SDP
Catalog No. 5000291952
Encompass_Preferred
Recombinant hepatocyte growth factor (HGF) derived from cynomolgus macaque and expressed in HEK293 cells. Supplied as a tag-free, lyophilized protein formulated for stability and intended for in vitro research use; acts as a ligand for the MET receptor and supports studies of signaling, proliferation, and receptor binding.
  • High purity: >95.0% as determined by reducing SDS-PAGE.
  • Expressed in HEK293 cells for eukaryotic post-translational modifications.
  • Tag free to preserve native activity and receptor interactions.
  • Formulated with trehalose, mannitol, and Tween 80 for stability.
  • Biologically active with ED50 1-10 ng/mL in cell-based assays.
  • Lyophilized for long-term storage; reconstitute to ≥100 μg/mL as recommended.
Catalog No. 50-002-91952 Supplier Medchemexpress LLC Supplier No. HYP75809100UG
Recombinant hepatocyte growth factor (HGF) derived from cynomolgus macaque and expressed in HEK293 cells. Supplied as a tag-free, lyophilized protein formulated for stability and intended for in vitro research use; acts as a ligand for the MET receptor and supports studies of signaling, proliferation, and receptor binding.
  • High purity: >95.0% as determined by reducing SDS-PAGE.
  • Expressed in HEK293 cells for eukaryotic post-translational modifications.
  • Tag free to preserve native activity and receptor interactions.
  • Formulated with trehalose, mannitol, and Tween 80 for stability.
  • Biologically active with ED50 1-10 ng/mL in cell-based assays.
  • Lyophilized for long-term storage; reconstitute to ≥100 μg/mL as recommended.