Learn More
Cusabio Technology LLC hepatocyte growth factor HGF
Catalog No. 5000049200
Encompass_Preferred
The Pig Hepatocyte Growth Factor (HGF) ELISA Kit is designed to quantitatively measure HGF levels in serum plasma and tissue homogenates using a sandwich assay format This kit detects HGF within a broad detection range of 62 5 pg/mL to 4000 pg/mL with a sensitivity of 15 6 pg/mL ensuring precise measurement of low-concentration samples The assay requires a short incubation period of 1-5 hours and utilizes 50-100 L of sample per test Results are determined by absorbance at 450 nm providing reliable and reproducible data for research applications Suitable for both routine analysis and specialized studies this kit offers a robust tool for evaluating HGF dynamics in porcine biological samples while maintaining high specificity and accuracy
Catalog No. 50-000-49200 Supplier Cusabio Technology LLC Supplier No. CSBE06795P
The Pig Hepatocyte Growth Factor (HGF) ELISA Kit is designed to quantitatively measure HGF levels in serum plasma and tissue homogenates using a sandwich assay format This kit detects HGF within a broad detection range of 62 5 pg/mL to 4000 pg/mL with a sensitivity of 15 6 pg/mL ensuring precise measurement of low-concentration samples The assay requires a short incubation period of 1-5 hours and utilizes 50-100 L of sample per test Results are determined by absorbance at 450 nm providing reliable and reproducible data for research applications Suitable for both routine analysis and specialized studies this kit offers a robust tool for evaluating HGF dynamics in porcine biological samples while maintaining high specificity and accuracy