Learn More
Worthington Biochemical Corporation HEPATOCYTE ISOLATION
SDP
Catalog No. NC0390085
Encompass
A component of the Hepatocyte and Papain Dissociation Systems. This material is 0.22 micron membrane filtered and lyophilized in autoclaved vials. A vial reconstituted with 0.5 ml of EBSS or equivalent yields a solution of 2000 units/ml of deoxyribonuclease.
Catalog No. NC0390085 Supplier Worthington Biochemical Corporation Supplier No. LK003170
A component of the Hepatocyte and Papain Dissociation Systems. This material is 0.22 micron membrane filtered and lyophilized in autoclaved vials. A vial reconstituted with 0.5 ml of EBSS or equivalent yields a solution of 2000 units/ml of deoxyribonuclease.