Learn More
Lonza Walkersville MM250 Hepatocyte Maintenance Medium
Catalog No. 501060963
Encompass
Serum-free hepatocyte maintenance medium for optimal performance of primary hepatocytes in monolayer or sandwich culture formats.This maintenance medium is specifically required for mouse plateable hepatocyte products.Benefits:Optimized for Hepatocyte Cell healthApplications:ADMEToxContent:250mLs basal medium 2.5mL supplement
Catalog No. 50-106-0963 Supplier Lonza Walkersville Supplier No. MM250
Serum-free hepatocyte maintenance medium for optimal performance of primary hepatocytes in monolayer or sandwich culture formats.This maintenance medium is specifically required for mouse plateable hepatocyte products.Benefits:Optimized for Hepatocyte Cell healthApplications:ADMEToxContent:250mLs basal medium 2.5mL supplement