Learn More
Enzo Life Sciences SCREEN-WELL Hepatotoxicity library (100µl/well)
SDP
Catalog No. 502010221
Encompass_Preferred
The SCREEN-WELL Hepatotoxicity Library is a focused collection of 238 compounds with defined and diverse hepatotoxicity, including steatosis, mitochondrial toxicity, Mallory body formation, cholestatic effects, and many more. A variety of structurally and mechanistically different compound classes are included, as well as nontoxic controls. The compounds are dissolved in DMSO at 10mM and aliquoted into deep-well plates. The library is an essential tool for predictive toxicology screening and assay development. Long Term Storage: -80°C. Applications: HTS .
Catalog No. 50-201-0221 Supplier Enzo Life Sciences Supplier No. BML28510100
The SCREEN-WELL Hepatotoxicity Library is a focused collection of 238 compounds with defined and diverse hepatotoxicity, including steatosis, mitochondrial toxicity, Mallory body formation, cholestatic effects, and many more. A variety of structurally and mechanistically different compound classes are included, as well as nontoxic controls. The compounds are dissolved in DMSO at 10mM and aliquoted into deep-well plates. The library is an essential tool for predictive toxicology screening and assay development. Long Term Storage: -80°C. Applications: HTS .