Learn More
ATCC HepatoXcell™ Primary Hepatocyte Maintenance Medium 1x
Catalog No. 5000549745
Encompass
HepatoXcell™ Maintenance Media is a complete, ready-to-use, phenol red free media formulated for the purpose of culturing fresh or cryopreserved primary hepatocytes. The animal by-product (ABP) free maintenance media has been optimally designed for primary hepatocyte culture and assays.
Catalog No. 50-005-49745 Supplier ATCC Supplier No. PCS450034
HepatoXcell™ Maintenance Media is a complete, ready-to-use, phenol red free media formulated for the purpose of culturing fresh or cryopreserved primary hepatocytes. The animal by-product (ABP) free maintenance media has been optimally designed for primary hepatocyte culture and assays.