Learn More
ATCC HepatoXcell™ Primary Hepatocyte Plating Medium 1x
Catalog No. 5000549961
Encompass
HepatoXcell™ Plating Media is a complete, ready-to-use, phenol red free media formulated for the purpose of plating fresh or cryopreserved primary hepatocytes. The plating media containing prequalified fetal bovine serum (FBS) has been optimally designed for primary hepatocyte seeding and successful attachment to collagen I coated plates.
Catalog No. 50-005-49961 Supplier ATCC Supplier No. PCS450038
HepatoXcell™ Plating Media is a complete, ready-to-use, phenol red free media formulated for the purpose of plating fresh or cryopreserved primary hepatocytes. The plating media containing prequalified fetal bovine serum (FBS) has been optimally designed for primary hepatocyte seeding and successful attachment to collagen I coated plates.