Learn More
ATCC HepatoXcell™ Primary Hepatocyte Thawing Medium 1x
Catalog No. 5000550166
Encompass
HepatoXcell™ Thawing Media is a ready-to-use, animal by-product (ABP) free solution, formulated to separate the live hepatocytes from the dead cells, non-hepatic cells as well as cell debris. The phenol-red free HepatoXcell Thawing Media also ensures cell quality is maintained during the thawing process as the cells recover from freezing.
Catalog No. 50-005-50166 Supplier ATCC Supplier No. PCS450032
HepatoXcell™ Thawing Media is a ready-to-use, animal by-product (ABP) free solution, formulated to separate the live hepatocytes from the dead cells, non-hepatic cells as well as cell debris. The phenol-red free HepatoXcell Thawing Media also ensures cell quality is maintained during the thawing process as the cells recover from freezing.