Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bioworld HEPBS Buffer 0.2M, pH 9.0, 100 mL

Catalog No. NC2212593
Encompass_Preferred

liquid, biotechnology grade, 0.2m, ph 9

This item is not returnable. View return policy
Catalog No. NC2212593 Supplier Bioworld Supplier No. 408200031
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

liquid, biotechnology grade, 0.2m, ph 9

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.