Learn More
Bachem Hepcidin-20 (human) Trifluoroacetate
SDP
Supplier:  Bachem 4065374.0500
Encompass_Preferred
Hepcidin-20 (human) Trifluoroacetate, H-Ile-Cys-Ile-Phe-Cys-Cys-Gly-Cys-Cys-His-Arg-Ser-Lys-Cys-Gly-Met-Cys-Cys-Lys-Thr-OH, 342790-23-2, C85H135N27O23S9, Mr 2191.77, 0.5mg. Hepcidin-20, a degradation product of hepcidin-25 found in plasma and urine, shows an increased antimicrobial activity compared to the full-length peptide, but it is not involved in iron metabolism.
Catalog No. 50-194-6660