Learn More
Carbosynth LLC. Hepcidin-24 (Human), 0.5mg, Disulfide bonds: Cys6- Cys22, Cys9-Cys12, Cys10- Cys18, and Cys13-Cys21, MW: (M.W. 2674.31), Molecular formula: C109H165N33O28S9
SDP
Supplier:  Carbosynth LLC. PLP3779PI0.5MG
Encompass_Preferred
Synonyms: H-Thr-His-Phe-Pro-Ile-Cys-Ile-Phe-Cys-Cys-Gly-Cys-Cys-His-Arg-Ser-Lys-Cys-Gly-Met-Cys-Cys-Lys-Thr-OH; des-Asp1-Hepcidin-24, THFPICIFCCGCCHRSKCGMCCKT, Application: An Internal Standard for Hepcidin Assays, Salt Form:, Trifluoroacetate Salt, Storage Temperature: -20°C, References: D.W. Swinkels, et al., PLoS ONE, 3: e2706 (2008).
Catalog No. 50-168-6608