Learn More
Bachem Hepcidin-24 (human) Trifluoroacetate
SDP
Supplier:  Bachem 4084346.0500
Encompass_Preferred
Hepcidin-24 (human) Trifluoroacetate, H-Thr-His-Phe-Pro-Ile-Cys-Ile-Phe-Cys-Cys-Gly-Cys-Cys-His-Arg-Ser-Lys-Cys-Gly-Met-Cys-Cys-Lys-Thr-OH, , C109H165N33O28S9, Mr 2674.31, 0.5mg. Hepcidin-24 is an excellent alternative to biotinylated or heavy isotope-labeled hepcidin-25 used as internal reference in the determination of Hep-25 by mass spectrometry-based methods, e.g. in human serum or urine.
Catalog No. 50-194-6663