Learn More
Bachem Hepcidin-25 (human) Trifluoroacetate
SDP
Supplier:  Bachem 4040671.0100
Encompass_Preferred
Hepcidin-25 (human) Trifluoroacetate, H-Asp-Thr-His-Phe-Pro-Ile-Cys-Ile-Phe-Cys-Cys-Gly-Cys-Cys-His-Arg-Ser-Lys-Cys-Gly-Met-Cys-Cys-Lys-Thr-OH, 1356390-47-0, C113H170N34O31S9, Mr 2789.40, 0.1mg. LEAP-1 (human), Hepc25 (human), Liver-Expressed Antimicrobial Peptide (human), PLTR (human), Putative Liver Tumor Regressor (human). Hepcidin-25 (hepatic bactericidal protein), also referred to as LEAP-1 (liver-expressed antimicrobial peptide) has been independently discovered by two groups in human blood ultrafiltrate and urine. The peptide which is predominantly expressed in the liver, belongs to a new vertebrate family of small antimicrobial peptides that contain 8 cysteine residues and show significant antibacterial and antifungal activity.
Catalog No. 50-194-6665