Learn More
Drg International Inc DRG Hepcidin-25 (Bioactive) HS ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA5782R
Encompass_Preferred
A high sensitive enzyme immunoassay for the quantitative measurement of Hepcidin-25 in serum or plasma (EDTA-, heparin- or citrate plasma). This product is for Research Use Only. The DRG Hepcidin 25 (bioactive) HS ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) based on the principle of competive binding. The microtiter wells are coated with a monoclonal (mouse) antibody directed towards an antigenic site of the Hepcidin-25 molecule.
Catalog No. 50-187-1235