Learn More
Bioss Hepcidin-25 Monoclonal Antibod
SDP
Catalog No. 5000064668
Encompass_Preferred
Hepcidin-25 Primary Antibody Unconjugated Host Species Rabbit Target Species Human Rat Clonality Monoclonal Applications WB RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol
Catalog No. 50-000-64668 Supplier Bioss Supplier No. BSM61456R
Hepcidin-25 Primary Antibody Unconjugated Host Species Rabbit Target Species Human Rat Clonality Monoclonal Applications WB RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol