Learn More
Carbosynth LLC. Hepcidin (Baboon, Cynomolgus, Macaque, and Gorilla), 1mg, Disulfide bonds: Cys7- Cys23, Cys10- Cys13, Cys11- Cys19, and Cys14- Cys22, MW: -2817.41, Molecular formula: C113H170N36O31S9
SDP
Supplier:  Carbosynth LLC. PLP3745PI1MG
Encompass_Preferred
Synonyms: H-Asp-Thr-His-Phe-Pro-Ile-Cys-Ile-Phe-Cys-Cys-Gly-Cys-Cys-His-Arg-Ser-Lys-Cys-Gly-Met-Cys-Cys-Arg-Thr-OH, Storage Temperature: -20°C, References: G.M. Morrison, et al., Mol Biol Evol., 20, 460 (2003).
Catalog No. 50-168-6596