Learn More
Carbosynth LLC. Hepcidin / LEAP-1 (Human) (0.1mg vial), 1ea, Disulfide bonds: Disulfide bonds between Cys7-Cys23, Cys10-Cys13, Cys11-Cys19, Cys14-Cys22, MW: 2789.4, Molecular formula: C113H170N34O31S9, CAS# 342790-21-0
SDP
Catalog No. 501686628
Encompass_Preferred
Synonyms: Asp-Thr-His-Phe-Pro-Ile-Cys-Ile-Phe-Cys-Cys-Gly-Cys-Cys-His-Arg-Ser-Lys-Cys-Gly-Met-Cys-Cys-Lys-Thr, Hepcidin / LEAP-1 (Human) (0.1mg vial), Hepcidin-25, DTHFPICIFCCGCCHRSKCGMCCKT, Application: Liver-Expressed Antimicrobial Peptide 1 (Human), Liver-Specific Antimicrobial Peptide / Iron-Regulatory Hormone, Storage Temperature: -20°C, References: M. Zasloff, Nature, 475, 389 (2002). (Review), A. Krause, et al., FEBS Lett., 480, 147 (2000). (Original; LEAP-1), C.H. Park, E.V. Valore, A.J. Waring, and T. Ganz, J. Biol. Chem., 276, 7806 (2001). (Original; Hepcidin)
Catalog No. 50-168-6628 Supplier Carbosynth LLC. Supplier No. PLP4392S0.1MG
Synonyms: Asp-Thr-His-Phe-Pro-Ile-Cys-Ile-Phe-Cys-Cys-Gly-Cys-Cys-His-Arg-Ser-Lys-Cys-Gly-Met-Cys-Cys-Lys-Thr, Hepcidin / LEAP-1 (Human) (0.1mg vial), Hepcidin-25, DTHFPICIFCCGCCHRSKCGMCCKT, Application: Liver-Expressed Antimicrobial Peptide 1 (Human), Liver-Specific Antimicrobial Peptide / Iron-Regulatory Hormone, Storage Temperature: -20°C, References: M. Zasloff, Nature, 475, 389 (2002). (Review), A. Krause, et al., FEBS Lett., 480, 147 (2000). (Original; LEAP-1), C.H. Park, E.V. Valore, A.J. Waring, and T. Ganz, J. Biol. Chem., 276, 7806 (2001). (Original; Hepcidin)