Learn More
Supelco Inc HEPES, 7365-45-9, MFCD00006158, 100 G
Catalog No. NC0855813
Encompass
Empirical Formula: (Hill Notation) C8H18N2O4S, Molecular Weight: 238.30, Synonym: 4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid), assay : ≥99.5% (titration). HEPES has been described as one of the best all-purpose buffers available for biological research. At biological pH, the molecule is zwitterionic, and is effective as a buffer at pH 6.8 to 8.2. HEPES has been used in a wide variety of applications, including tissue culture. It is commonly used to buffer cell culture media in air. HEPES finds its usage in in vitro experiments on Mg.
Catalog No. NC0855813 Supplier Supelco Inc Supplier No. H3375100G
Empirical Formula: (Hill Notation) C8H18N2O4S, Molecular Weight: 238.30, Synonym: 4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid), assay : ≥99.5% (titration). HEPES has been described as one of the best all-purpose buffers available for biological research. At biological pH, the molecule is zwitterionic, and is effective as a buffer at pH 6.8 to 8.2. HEPES has been used in a wide variety of applications, including tissue culture. It is commonly used to buffer cell culture media in air. HEPES finds its usage in in vitro experiments on Mg.