Learn More
Dojindo Molecular Technologies Inc HEPES (100 G), Good's buffer, CAS: 7365-45-9, Purity: ≥99.0% (Titration), pKa=7.55, Active pH 6.8-8.2, made in Japan
SDP
Catalog No. NC1808672
Encompass_Preferred
Buffers perform an important task in biological experiments, often designed to keep pH within the 6-8 range where most biological reactions occur. Dr. Norman Good, the developer of HEPES, wanted to create buffers that prioritized this function, while also providing other benefits. Dr. Good produced a series of “Good buffers” – including HEPES (2-[4-(2-Hydroxyethyl)-1-piperazinyl]ethanesulfonic acid) – that are nearly ubiquitous in their usage even now. The Good series of buffers share the following additional characteristics: 1) High water-solubility. 2) Low cell membrane permeability. 3) Consistent acid-base dissociation constants. 4) Low metal-chelating capability. 5) High chemical stability. 6)Low absorption spectra in UV and visible regions.
Catalog No. NC1808672 Supplier Dojindo Molecular Technologies Inc Supplier No. GB1012
Add to Cart
Add to Cart
Buffers perform an important task in biological experiments, often designed to keep pH within the 6-8 range where most biological reactions occur. Dr. Norman Good, the developer of HEPES, wanted to create buffers that prioritized this function, while also providing other benefits. Dr. Good produced a series of “Good buffers” – including HEPES (2-[4-(2-Hydroxyethyl)-1-piperazinyl]ethanesulfonic acid) – that are nearly ubiquitous in their usage even now. The Good series of buffers share the following additional characteristics: 1) High water-solubility. 2) Low cell membrane permeability. 3) Consistent acid-base dissociation constants. 4) Low metal-chelating capability. 5) High chemical stability. 6)Low absorption spectra in UV and visible regions.