Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEPES (Fine White Crystals/Molecular Biology), Fisher BioReagents™

Catalog No. BP310100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
Glass Bottle
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP310-100 100 g Glass Bottle
BP310-500 500 g Poly Bottle
BP310-1 1 kg Poly Bottle
BP310-5 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. BP310-100
  • $245.00
    $168.65 / Each

    Save $76.35

    31% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $1,151.00
    $793.65 / Case of 6 EA

    Save $357.35

    31% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Commonly used buffering agent | CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol

Commonly used buffering agent

HEPES is commonly used as a buffering agent.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HEPES
Absorbance 0.01 max. (0.1M solution) at 280nm
CAS 7365-45-9
Color White
pH 5.0 to 6.5
Physical Form Powder or Solid
Quantity 100 g
DNase DNase free
Purity Grade Notes DNase-, RNase- and Protease-Free
Assay Percent Range ≥99 %
Identification Pass Test
Molecular Formula C8H18N2O4S
MDL Number MFCD00006158
Packaging Glass Bottle
Protease Protease free
Merck Index 15, 4689
Synonym N-(2-Hydroxyethyl)piperazine-N'-2-ethanesulfonic Acid
Solubility Information Soluble in water
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
Molecular Weight (g/mol) 238.30
PubChem CID 23831
ChEBI CHEBI:42334
Percent Purity ≥99%
Grade Molecular Biology
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.