Learn More
Apexbio Technology LLC HEPES(Synonyms: HEPES buffer, 4-(2-Hydroxyethyl)-1-piperazineethanesulfonic acid), 250g, CAS: 7365-45-9.
SDP
Catalog No. 502240295
Encompass_Preferred
HEPES (CAS 7365-45-9) is a zwitterionic sulfonic acid buffer commonly utilized in biomedical research due to its capacity to maintain physiological pH stability within a buffering range of approximately pH 6 8 to 8 2 Mechanistically HEPES modulates intracellular pH balance thereby promoting lysosomal biogenesis and supporting associated cellular processes It is routinely employed in cell culture systems to precisely regulate environmental conditions and is relevant in biochemical assays involving specific cellular signaling pathways and intracellular pH-dependent phenomena
This item is not returnable. View return policy
Catalog No. 50-224-0295 Supplier Apexbio Technology LLC Supplier No. B7302250000
HEPES (CAS 7365-45-9) is a zwitterionic sulfonic acid buffer commonly utilized in biomedical research due to its capacity to maintain physiological pH stability within a buffering range of approximately pH 6 8 to 8 2 Mechanistically HEPES modulates intracellular pH balance thereby promoting lysosomal biogenesis and supporting associated cellular processes It is routinely employed in cell culture systems to precisely regulate environmental conditions and is relevant in biochemical assays involving specific cellular signaling pathways and intracellular pH-dependent phenomena