Learn More
Selleck Chemical LLC HEPES
SDP
Catalog No. 5000002844
Encompass_Preferred
HEPES is a nonvolatile zwitterionic chemical buffering agent which is broadly applied in cell culture HEPES is effective at pH 6 8 to 8 2 HEPES is also a potent inducer of lysosome biogenesis
Catalog No. 50-000-02844 Supplier Selleck Chemical LLC Supplier No. S683125MG
HEPES is a nonvolatile zwitterionic chemical buffering agent which is broadly applied in cell culture HEPES is effective at pH 6 8 to 8 2 HEPES is also a potent inducer of lysosome biogenesis