Learn More
Sigma Aldrich Fine Chemicals Biosciences HEPES, 7365-45-9, MFCD00006158, 100g
Catalog No. 501657163
Encompass
Empirical Formula (Hill Notation) C8H18N2O4S, ≥99.5% (titration), Molecular Weight 238.30, Synonym: 4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid)
Catalog No. 50-165-7163 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H3375100G
Empirical Formula (Hill Notation) C8H18N2O4S, ≥99.5% (titration), Molecular Weight 238.30, Synonym: 4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid)