Learn More
Medchemexpress LLC HY-D0857 500mg Medchemexpress, HEPES CAS:7365-45-9 Purity:>98%
SDP
Catalog No. 502031273
Encompass_Preferred
Medchemexpress, HY-D0857 500mg HEPES CAS:7365-45-9 HEPES, a zwitterionic chemical buffering agent, is broadly applied in cell culture. HEPES is effective at pH 6.8 to 8.2. HEPES is a potent inducer of lysosome biogenesis[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1273 Supplier Medchemexpress LLC Supplier No. HYD0857500MG
Medchemexpress, HY-D0857 500mg HEPES CAS:7365-45-9 HEPES, a zwitterionic chemical buffering agent, is broadly applied in cell culture. HEPES is effective at pH 6.8 to 8.2. HEPES is a potent inducer of lysosome biogenesis[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.