Learn More
Sigma Aldrich Fine Chemicals Biosciences HEPES 99.5 titration25KG
Catalog No. 502914431
Encompass
HEPES has been described as one of the best all-purpose buffers available for biological research. At biological pH the molecule is zwitterionic and is effective as a buffer at pH 6.8 to 8.2. HEPES has been used in a wide variety of applications including tissue culture. It is commonly used to buffer cell culture media in air. HEPES finds its usage in in vitro experiments on Mg.
Catalog No. 50-291-4431 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H337525KG
Add to Cart
Add to Cart
Request a Quote
HEPES has been described as one of the best all-purpose buffers available for biological research. At biological pH the molecule is zwitterionic and is effective as a buffer at pH 6.8 to 8.2. HEPES has been used in a wide variety of applications including tissue culture. It is commonly used to buffer cell culture media in air. HEPES finds its usage in in vitro experiments on Mg.