Learn More
Teknova HEPES Assay Diluent, pH 7.5. 1000mL, Sterile. 2 pack.
SDP
Catalog No. 502387426
Encompass
0.59% HEPES Free Acid,0.65% HEPES Sodium Salt,0.90% Sodium Chloride,0.05% Tween-20,1.00% Bovine Serum Albumin (BSA)
Catalog No. 50-238-7426 Supplier Teknova Supplier No. 2A30302PK
0.59% HEPES Free Acid,0.65% HEPES Sodium Salt,0.90% Sodium Chloride,0.05% Tween-20,1.00% Bovine Serum Albumin (BSA)