Learn More
Boston Bioproducts Inc HEPES Buffer, 1 M, pH 7.5, 500 mL
Catalog No. NC1463291
Encompass
This product is a 1 M HEPES Buffer solution, pH 7.5, which is essential for maintaining stable physiological pH in various biological applications. It is rigorously quality-controlled and specifically formulated for precision in sensitive biochemical and molecular biology research. The high-purity zwitterionic buffering agent is ready-to-use and suitable for diverse applications such as cell culture, protein purification, and live cell imaging.
  • Stable pH control maintaining 6.8 to 8.2
  • Broad applicability in various research areas
  • Convenient format as a ready-to-use solution
  • High purity, formulated with USP Compendial-Grade raw materials
  • CO₂-independent buffering for reliable pH stability
  • Filtered through a 0.22-micron filter for enhanced purity
Catalog No. NC1463291 Supplier Boston Bioproducts Inc Supplier No. BBH75K500ML
This product is a 1 M HEPES Buffer solution, pH 7.5, which is essential for maintaining stable physiological pH in various biological applications. It is rigorously quality-controlled and specifically formulated for precision in sensitive biochemical and molecular biology research. The high-purity zwitterionic buffering agent is ready-to-use and suitable for diverse applications such as cell culture, protein purification, and live cell imaging.
  • Stable pH control maintaining 6.8 to 8.2
  • Broad applicability in various research areas
  • Convenient format as a ready-to-use solution
  • High purity, formulated with USP Compendial-Grade raw materials
  • CO₂-independent buffering for reliable pH stability
  • Filtered through a 0.22-micron filter for enhanced purity