Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEPES, Biological Buffer, 99%, Spectrum™ Chemical
SDP

Catalog No. 18610778 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
500 g
1 kg
5 kg
12 kg
Packaging:
Poly Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-610-778 25 g Poly Bottle
18-610-776 100 g Poly Bottle
18-610-779 250 g Poly Bottle
18-610-781 500 g Poly Bottle
18-610-775 1 kg Poly Bottle
18-610-780 5 kg Poly Bottle
18-610-777 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-610-778 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. HE15525GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1

Specifications

CAS Max % 1
For Use With (Application) Pharma and Biopharma
Physical Form Crystalline
Quantity 25 g
Loss on Drying 0.005
Packaging Poly Bottle
Percent Purity 99%
Grade Reagent
Chemical Name or Material 4-(2-Hydroxyethyl)-1-Piperazineethanesulfonic Acid

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.