Learn More
Sigma Aldrich Fine Chemicals Biosciences HEPES BioPerformance Certified, ≥99.5% (titration), suitable for cell culture | 7365-45-9 | MFCD00006158 | 500g
Catalog No. NC0813666
Encompass
HEPES BioPerformance Certified, ≥99.5% (titration), suitable for cell culture | Purity: | MW: 238.3 | 7365-45-9 | MFCD00006158 | 500g
Catalog No. NC0813666 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H4034500G
HEPES BioPerformance Certified, ≥99.5% (titration), suitable for cell culture | Purity: | MW: 238.3 | 7365-45-9 | MFCD00006158 | 500g