Learn More
Sigma Aldrich Fine Chemicals Biosciences HEPES BioUltra for molecul50G
Catalog No. 502898123
Encompass
HEPES has been described as one of the best all-purpose buffers available for biological research. At biological pH the molecule is zwitterionic and is effective as a buffer at pH 6.8 to 8.2. HEPES has been used in a wide variety of applications including tissue culture. It is commonly used to buffer cell culture media in air. HEPES finds its usage in in vitro experiments on Mg.
Catalog No. 50-289-8123 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 5445750GF
HEPES has been described as one of the best all-purpose buffers available for biological research. At biological pH the molecule is zwitterionic and is effective as a buffer at pH 6.8 to 8.2. HEPES has been used in a wide variety of applications including tissue culture. It is commonly used to buffer cell culture media in air. HEPES finds its usage in in vitro experiments on Mg.