Learn More
Sigma Aldrich Fine Chemicals Biosciences HEPES BioXtra suitable for500G
Catalog No. 502907118
Encompass
HEPES has been described as one of the best all-purpose buffers available for biological research. At biological pH the molecule is zwitterionic and is effective as a buffer at pH 6.8 to 8.2 (pKa 7.55). HEPES has been used in a wide variety of applications including tissue culture. It is commonly used to buffer cell culture media in air. HEPES finds its usage in in vitro experiments on Mg. It is typically used in cell culture at concentration between 5mM to 30mM.
Catalog No. 50-290-7118 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H6147500G
HEPES has been described as one of the best all-purpose buffers available for biological research. At biological pH the molecule is zwitterionic and is effective as a buffer at pH 6.8 to 8.2 (pKa 7.55). HEPES has been used in a wide variety of applications including tissue culture. It is commonly used to buffer cell culture media in air. HEPES finds its usage in in vitro experiments on Mg. It is typically used in cell culture at concentration between 5mM to 30mM.