Learn More
Promocell GmbH PromoCell Hepes BSS (30 mM) 125 ml, HEPES Buffered Saline Solution
Catalog No. 50306361
Encompass
Buffers and Salt Solutions are frequently used in tissue culture, cell isolation procedures, and molecular biology.For convenience, PromoCell offers ready-to-use Buffers and Salt Solutions. They are prepared from raw materials of the highest quality using tissue culture grade water.HEPES Buffered Saline Solution (30 mM).
Catalog No. 50-306-361 Supplier Promocell GmbH Supplier No. C40010
Buffers and Salt Solutions are frequently used in tissue culture, cell isolation procedures, and molecular biology.For convenience, PromoCell offers ready-to-use Buffers and Salt Solutions. They are prepared from raw materials of the highest quality using tissue culture grade water.HEPES Buffered Saline Solution (30 mM).