Learn More
Promocell GmbH PromoCell Hepes BSS (30 mM) 250 ml, HEPES Buffered Saline Solution
Catalog No. 50306362
Encompass
Buffers and Salt Solutions are frequently used in tissue culture, cell isolation procedures, and molecular biology.For convenience, PromoCell offers ready-to-use Buffers and Salt Solutions. They are prepared from raw materials of the highest quality using tissue culture grade water.HEPES Buffered Saline Solution (30 mM).
Catalog No. 50-306-362 Supplier Promocell GmbH Supplier No. C40020
Add to Cart
Add to Cart
Buffers and Salt Solutions are frequently used in tissue culture, cell isolation procedures, and molecular biology.For convenience, PromoCell offers ready-to-use Buffers and Salt Solutions. They are prepared from raw materials of the highest quality using tissue culture grade water.HEPES Buffered Saline Solution (30 mM).