Learn More
Lonza Walkersville HEPES Buffered Saline Solution - 100 ml
Catalog No. NC9552375
Encompass
HEPES-Buffered Saline Solution (BSS), is used to neutralize the complex proteins in growth medium that may inactivate trypsin during trypsinization.HEPES-BSS is a component of the ReagentPack Subculture kit (catalog number CC-5034). It is also available in a 500 mL size (catalog number CC-5024).Content:1 x Bottle HEPES Buffered Solution 100 mL
Catalog No. NC9552375 Supplier Lonza Walkersville Supplier No. CC5022
HEPES-Buffered Saline Solution (BSS), is used to neutralize the complex proteins in growth medium that may inactivate trypsin during trypsinization.HEPES-BSS is a component of the ReagentPack Subculture kit (catalog number CC-5034). It is also available in a 500 mL size (catalog number CC-5024).Content:1 x Bottle HEPES Buffered Solution 100 mL