Learn More
Biotang Inc HEPES Buffered Saline 5X, pH 6.5. 125 mM HEPES, 750 mM Sodium Chloride, pH 6.5. Prepared in 18.2 megohms water and filtered through 0.22 micron filter. BB-545
Catalog No. 50155804 Shop All BIOTANG Inc Products
Encompass_Preferred
HEPES Buffered Saline 5X, pH 6.5. 125 mM HEPES, 750 mM Sodium Chloride, pH 6.5. Prepared in 18.2 megohms water and filtered through 0.22 micron filter. BB-545.
Catalog No. 50-155-804 Supplier Biotang Inc Supplier No. BTBB455
HEPES Buffered Saline 5X, pH 6.5. 125 mM HEPES, 750 mM Sodium Chloride, pH 6.5. Prepared in 18.2 megohms water and filtered through 0.22 micron filter. BB-545.