Learn More
Boston Bioproducts Inc HEPES Buffer (0.5 M, pH 7.4, pH Adjusted with KOH) - 250ml
Catalog No. NC0686037
Encompass
The HEPES [4-(2-hydroxyethyl)-1-piperazineethanesulfonic acid)] Buffer is a versatile Good's zwitterionic buffering agent widely used in various applications in biochemistry and molecular biology. It maintains a stable pH in the range of 6.8 to 8.2, making it ideal for various applications at physiological pH, including cell culture, protein purification, isoelectric focusing, enzyme studies, bioprocessing, and live cell imaging. This formulation is adjusted with KOH.
Catalog No. NC0686037 Supplier Boston Bioproducts Inc Supplier No. BB2076K250ML
The HEPES [4-(2-hydroxyethyl)-1-piperazineethanesulfonic acid)] Buffer is a versatile Good's zwitterionic buffering agent widely used in various applications in biochemistry and molecular biology. It maintains a stable pH in the range of 6.8 to 8.2, making it ideal for various applications at physiological pH, including cell culture, protein purification, isoelectric focusing, enzyme studies, bioprocessing, and live cell imaging. This formulation is adjusted with KOH.