Learn More
Axxora LLC Hepes Buffer 1 M pH 7.5
SDP
Catalog No. NC1776877
Encompass
Hepes Buffer is one of the best general purpose buffers available for biological research. It has been used to advantage in tissue culture, oxidative phosphorylation and protein synthesis with cell-free bacterial systems. Hepes is a recommended buffer for the glutamate binding assay, prevents binding to non-receptor materials.
Catalog No. NC1776877 Supplier Axxora LLC Supplier No. JBSBU10675
Hepes Buffer is one of the best general purpose buffers available for biological research. It has been used to advantage in tissue culture, oxidative phosphorylation and protein synthesis with cell-free bacterial systems. Hepes is a recommended buffer for the glutamate binding assay, prevents binding to non-receptor materials.