Learn More
ILEX LIFE SCIENCES LLC PAN-Biotech HEPES Buffer 1M (100 ml)
SDP
Catalog No. 502388618
Encompass_Preferred
HEPES Buffer 1M. QUANTITY: 100 ml. MANUFACTURED BY: Pan-Biotech (Germany). STORAGE: 2 to 8 degrees Celsius. DESCRIPTION: HEPES (4-(2-hydroxyethyl)-1-piperazineethanesulfonic acid) is a zwitterionic sulfonic acid buffering agent; one of the twenty Good's buffers. HEPES is widely used in cell culture, largely because it is better at maintaining physiological pH despite changes in carbon dioxide concentration (produced by aerobic respiration) when compared to bicarbonate buffers, which are also commonly used in cell culture. COMPOSITION: HEPES 238.31 g/L.
Catalog No. 50-238-8618 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0501100
HEPES Buffer 1M. QUANTITY: 100 ml. MANUFACTURED BY: Pan-Biotech (Germany). STORAGE: 2 to 8 degrees Celsius. DESCRIPTION: HEPES (4-(2-hydroxyethyl)-1-piperazineethanesulfonic acid) is a zwitterionic sulfonic acid buffering agent; one of the twenty Good's buffers. HEPES is widely used in cell culture, largely because it is better at maintaining physiological pH despite changes in carbon dioxide concentration (produced by aerobic respiration) when compared to bicarbonate buffers, which are also commonly used in cell culture. COMPOSITION: HEPES 238.31 g/L.