Learn More
Boston Bioproducts Inc HEPES Buffer (1 M, pH 7.4, pH Adjusted with KOH) - 500ml
Catalog No. NC2146875
Encompass
The HEPES [4-(2-hydroxyethyl)-1-piperazineethanesulfonic acid)] Buffer is a versatile Good's zwitterionic buffering agent widely used in various applications in biochemistry and molecular biology. It maintains a stable pH in the range of 6.8 to 8.2, making it ideal for various applications at physiological pH, including cell culture, protein purification, isoelectric focusing, enzyme studies, bioprocessing, and live cell imaging. This buffer is adjusted with KOH.
This item is not returnable. View return policy
Catalog No. NC2146875 Supplier Boston Bioproducts Inc Supplier No. BBH74K500ML
The HEPES [4-(2-hydroxyethyl)-1-piperazineethanesulfonic acid)] Buffer is a versatile Good's zwitterionic buffering agent widely used in various applications in biochemistry and molecular biology. It maintains a stable pH in the range of 6.8 to 8.2, making it ideal for various applications at physiological pH, including cell culture, protein purification, isoelectric focusing, enzyme studies, bioprocessing, and live cell imaging. This buffer is adjusted with KOH.