Learn More
Electron Microscopy Sciences Hepes Buffer Solution 0.2M, 7.0-8.0 pH 500 ML
SDP
Catalog No. 50980222
Encompass_Preferred
Our buffers are ideal for maintaining a constant pH during fixation and have an ionic constitution to prevent the extraction of cellular components or the deposition of precipitates of fixative chemicals. The buffers are prepared fresh for each order and conveniently packaged. Desired pH should be specified when ordering.
pH: 7.0-8.0
Catalog No. 50-980-222 Supplier Electron Microscopy Sciences Supplier No. 11494
Add to Cart
Add to Cart
Our buffers are ideal for maintaining a constant pH during fixation and have an ionic constitution to prevent the extraction of cellular components or the deposition of precipitates of fixative chemicals. The buffers are prepared fresh for each order and conveniently packaged. Desired pH should be specified when ordering.
pH: 7.0-8.0