Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ EmbryoMax™ 1M HEPES Buffer Solution

Catalog No. tms003c Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TMS003C 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TMS003C Supplier MilliporeSigma™ Supplier No. TMS003C
Only null left
Add to Cart
Add to Cart

available in a 100mL format and may be used for routine mouse embryonic stem cell culture applications.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20° C for 3 years
For Use With (Application) Stem Cell Culture
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.