Learn More
Boston Bioproducts Inc Tyrode's Solution (with HEPES & 0.25% BSA, without Calcium) - 500ml
Catalog No. NC1277536
Encompass
Tyrode’s Solution is an isotonic buffer solution that plays a crucial role in cell culture and tissue preservation. It is formulated with inorganic salts, HEPES, BSA, and a carbohydrate energy source to provide cells with essential nutrients while maintaining a stable pH and osmotic balance. The balanced composition of this buffer maintains the structural and physiological integrity of cells during biological research and development. This product is made without calcium.
Catalog No. NC1277536 Supplier Boston Bioproducts Inc Supplier No. BSS365500ML
Tyrode’s Solution is an isotonic buffer solution that plays a crucial role in cell culture and tissue preservation. It is formulated with inorganic salts, HEPES, BSA, and a carbohydrate energy source to provide cells with essential nutrients while maintaining a stable pH and osmotic balance. The balanced composition of this buffer maintains the structural and physiological integrity of cells during biological research and development. This product is made without calcium.