Learn More
Research Products International Corp HEPES Buffered Saline Solution 2X, 500 Milliliters
SDP
Catalog No. 50997884
Encompass_Preferred
2X Formulation:
NaCI: 280 mM
KCI: 10 mM
Na2HPO4: 1.5 mM
Dextrose: 12 mM
HEPES: 50 mM
pH: 7.0
Catalog No. 50-997-884 Supplier Research Products International Corp Supplier No. H17100500.0
2X Formulation:
NaCI: 280 mM
KCI: 10 mM
Na2HPO4: 1.5 mM
Dextrose: 12 mM
HEPES: 50 mM
pH: 7.0