Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals HEPES-buffered saline (5X, pH 7.4), low endotoxin

Catalog No. AAJ67502AK
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67502AK 250 mL
AAJ67502AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ67502AK Supplier Thermo Scientific Chemicals Supplier No. J67502AK
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HEPES-buffered saline (5X, pH 7.4)
Name Note low endotoxin
Color Colorless
Physical Form Liquid
Quantity 250 mL
What are the concentrations of the components of the HEPES-buffered saline (5X, pH 7.4), low endotoxin?

The HEPES-buffered saline (5X, pH 7.4), low endotoxin contains 125 mM HEPES and 750 mM Sodium Chloride, prepared in 18.2 megohm-cm±1 low endotoxin water and filtered through a 0.22-micron filter.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.