Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEPES, cGMP Manufactured, 99.5%, Spectrum™ Chemical
SDP

Catalog No. 18603106 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
1 kg
12 kg
25 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-603-106 25 kg
18-603-104 1 kg
18-603-105 12 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-603-106 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. H180425KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

H1804, 7365-45-9, C8H18O4N2S

Spectrum™ Chemical HEPES, cGMP Manufactured is a zwitterionic biological buffer and is one of Good's buffers widely used in cell culture. Spectrum cGMP grade products that have been manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1

Specifications

CAS Max % 100%
For Use With (Application) Pharma and Biopharma
pH 5.4 to 5.6
Physical Form Crystalline Powder or Crystals
Quantity 25 kg
Solution Type Buffer
Packaging Poly Bottle
Percent Purity 99.5%
Grade Reagent
Chemical Name or Material 4-(2-Hydroxyethyl)-1-Piperazineethanesulfonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.